Learn More
Cole-Parmer Utility Funnel Assortment Pack, 3/pk; 5ml, 25ml, 40ml
Supplier:  Cole-Parmer 6310032
Encompass_Preferred
Polypropylene funnels have eight internal ribs to facilitate filtration. Autoclavable.
Catalog No. NC1077455