Learn More
Fine Science Tools Utility Scissors, Stainless Steel, Straight, 21 cm
SDP
Catalog No. NC9754305
Encompass
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 85mm/ Alloy / Material: Stainless Steel/ Length: 21cm/ Serrated: No
Catalog No. NC9754305 Supplier Fine Science Tools Supplier No. 3750000
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 85mm/ Alloy / Material: Stainless Steel/ Length: 21cm/ Serrated: No