Learn More
Sklar Instruments Econo Multi-Cut Utility Scissors, Sklar, Black, 5-1/2 in., Blunt/Blunt, Angled, Serrated, Finger Rings, Stainless Steel / Plastic. Econo Utility Scissors are a multipurpose instrument used for cutting non-sterile materials. Boxes of 12.
SDP
Catalog No. 501437523 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Multi-Cut Utility Scissors, Sklar, Black, 5-1/2 in., Blunt/Blunt, Angled, Serrated, Finger Rings, Stainless Steel / Plastic. Econo Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. These scissors feature plastic finger ring handles with one ring that is larger than the other. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is angled with one serrated and one smooth edge, blunt/blunt tips, and a length of 5-1/2 inches. They are sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7523 Supplier Sklar Instruments Supplier No. 21101
Econo Multi-Cut Utility Scissors, Sklar, Black, 5-1/2 in., Blunt/Blunt, Angled, Serrated, Finger Rings, Stainless Steel / Plastic. Econo Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. These scissors feature plastic finger ring handles with one ring that is larger than the other. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is angled with one serrated and one smooth edge, blunt/blunt tips, and a length of 5-1/2 inches. They are sold in boxes of 12.