Learn More
MSC Carlisle Floater Scrub Brush, 8 in, Polyester Bristle, Yellow
Catalog No. 502836655
Encompass
This floater scrub brush is designed for various food service and commercial kitchen applications. It features a durable plastic block and stiff polyester bristles, providing oil resistance and effective scrubbing. The 8-inch brush is color-coded, which can assist in HACCP compliance and prevent cross-contamination in food processing environments.
  • Floater scrub brush
  • Color coded for HACCP compliance
  • Stiff polyester bristles
  • Oil-resistant design
  • Durable plastic block
  • Suitable for foodservice, commercial kitchens, and food processing equipment
Catalog No. 50-283-6655 Supplier MSC Supplier No. 75636449
This floater scrub brush is designed for various food service and commercial kitchen applications. It features a durable plastic block and stiff polyester bristles, providing oil resistance and effective scrubbing. The 8-inch brush is color-coded, which can assist in HACCP compliance and prevent cross-contamination in food processing environments.
  • Floater scrub brush
  • Color coded for HACCP compliance
  • Stiff polyester bristles
  • Oil-resistant design
  • Durable plastic block
  • Suitable for foodservice, commercial kitchens, and food processing equipment