Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk
GREENER_CHOICE

Catalog No. 12111131 Shop All OHAUS Products
Change view
Click to view available options
Volume (Metric):
250 μL
1000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12-111-131 250 μL
12-111-137 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-111-131 Supplier BIOTIX™ Supplier No. 63300079
Get Extra Pipette Products with Purchase
View all promotions
Add to Cart
Add to Cart

Top quality pipette tips in resealable bags. Validated for fit and accuracy on universal pipettes.

  • Validated for fit and accuracy on standard pipettes like Eppendorf™, Gilson™ and Biohit™
  • Low binding for Protein, DNA and nanoparticles
  • Re-sealable bag
  • Tips must be racked by hand
  • Certifications

    ISO 9001

    TRUSTED_SUSTAINABILITY

    Specifications

    Description 250 μL, Bulk
    Volume (Metric) 250 μL
    Filtered Non-filtered
    Sterility Non-sterile
    Color Clear
    Autoclavable Not Autoclavable
    Model Universal
    Length (Metric) 59.54 mm
    Certifications/Compliance ISO 9001
    Clean Claims Fully Recyclable
    For Use With (Application) Low binding for Protein, DNA, and nanoparticles
    Format Bulk
    Tip Style Ergonomic
    Quantity 1000 tips/bag
    Product Type Pipette Tips
    Disposable Single-use
    For Use With (Equipment) Universal Fit
    Graduated Yes
    Product Line uTip
    Material Polypropylene
    Shape Conical
    Graduations 10 μL, 50 μL, 100 μL, 200 μL
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.