Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk
GSA_VAGREENER_CHOICE

Catalog No. 12111137 Shop All OHAUS Products
Change view
Click to view available options
Volume (Metric):
250 μL
1000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12-111-137 1000 μL
12-111-131 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-111-137 Supplier BIOTIX™ Supplier No. 63300081
Get Extra Pipette Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Top quality pipette tips in resealable bags. Validated for fit and accuracy on universal pipettes.

  • Validated for fit and accuracy on standard pipettes like Eppendorf™, Gilson™ and Biohit™
  • Low binding for Protein, DNA and nanoparticles
  • Re-sealable bag
  • Tips must be racked by hand
  • Certifications

    ISO 9001

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    TRUSTED_SUSTAINABILITY

    Specifications

    Description 1000 μL, Bulk
    Volume (Metric) 1000 μL
    Filtered Non-filtered
    Sterility Non-sterile
    Color Clear
    Autoclavable Not Autoclavable
    Model Universal
    Length (Metric) 81.82 mm
    Certifications/Compliance ISO 9001
    Clean Claims Fully Recyclable
    For Use With (Application) Low binding for Protein, DNA, and nanoparticles
    Format Bulk
    Tip Style Ergonomic
    Quantity 1000 tips/bag
    Product Type Pipette Tips
    Disposable Single-use
    For Use With (Equipment) Universal Fit
    Graduated Yes
    Product Line uTip
    Material Polypropylene
    Shape Conical
    Graduations 200 μL, 500 μL, 1000 μL
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.