Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ uTIP™ Non-Sterile Low Retention Universal Pipette Tips, Standard
GREENER_CHOICE

Catalog No. 12111128 Shop All OHAUS Products
Change view
Click to view available options
Volume (Metric):
250 μL
1000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12-111-128 250 μL
12-111-134 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-111-128 Supplier BIOTIX™ Supplier No. 63300058
Get Extra Pipette Products with Purchase
View all promotions
Add to Cart
Add to Cart

Designed with FlexFit™ to lower necessary insertion and ejection force by ≈70% to make secure seal. Non-Filter Pipette Tips for Universal Pipettes are manufactured with low retention X-Resin™ giving tighter CV values.

  • Compatible with the widest range of industry leading pipettes - Fits all standard pipettes
  • Accurate and precise channel-channel draw on multichannel pipettes
  • Secure seal on single and multichannel models
  • FlexFit™ seals on your pipette with less force, reducing your ejection force
  • FlexFit™ technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
  • X-Resin™ maximizes your sample recovery by creating a low binding surface for DNA, proteins, and nanoparticles - resulting in tighter CV values
  • Blade™ lowers chance of hanging droplet formation by optimizing surface area at distal end of tip - ensures a clean delivery of your sample, increasing accuracy and precision
  • Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
  • Better reprodicibility
  • Certifications

    ISO 9001

    TRUSTED_SUSTAINABILITY

    Specifications

    Description 250 μL, Racked
    Volume (Metric) 250 μL
    Filtered Non-filtered
    Sterility Non-sterile
    Color Clear
    Rack Color Green
    Autoclavable Autoclavable
    Model Universal
    Length (Metric) 59.54 mm
    Certifications/Compliance ISO 9001
    Clean Claims Reusable as a 96 well reservoir
    For Use With (Application) Low binding for Protein, DNA, and nanoparticles
    Format Hinged Rack
    Tip Style Ergonomic
    Packaging Racked
    Quantity 10 racks/pk
    Product Type Pipette Tips
    Disposable Single-use
    For Use With (Equipment) Universal Fit
    Graduated Yes
    Product Line uTip
    Material Polypropylene
    Shape Conical
    Graduations 10 μL, 50 μL, 100 μL, 200 μL
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.