Learn More
Apexbio Technology LLC UTP Solution (100 mM) 250uL
SDP
Catalog No. 502240945
Encompass_Preferred
UTP also known as Uridine-5 -triphosphate is applicable to various molecular biology-related applications such as in vitro transcription RNA amplification and siRNA synthesis UTP can also participate in galactose metabolism where the activated form UDP-galactose is converted into UDP-glucose subsequently entering the glycogen synthesis pathway This product is a colorless and transparent aqueous solution prepared from UTP trisodium salt with purity more than 99% (HPLC) concentration 100 mM and is free from DNase and RNase contamination
Catalog No. 50-224-0945 Supplier Apexbio Technology LLC Supplier No. K10480.25
UTP also known as Uridine-5 -triphosphate is applicable to various molecular biology-related applications such as in vitro transcription RNA amplification and siRNA synthesis UTP can also participate in galactose metabolism where the activated form UDP-galactose is converted into UDP-glucose subsequently entering the glycogen synthesis pathway This product is a colorless and transparent aqueous solution prepared from UTP trisodium salt with purity more than 99% (HPLC) concentration 100 mM and is free from DNase and RNase contamination