Learn More
Aviva Systems Biology Corp Utp15 Antibody - middle region (ARP60525_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60525_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Utp15. It was validated on Western Blot. . Alternate names: AW544865ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-073