Learn More
Aviva Systems Biology Corp UTP23 Antibody - C-terminal region (ARP60526_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60526_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UTP23. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human UTP23. Alternate names: C8orf53, MGC14595ApplicationWB. Species reactivity: Cow, Dog, Human, Pig, Rabbit
Catalog No. 50-287-074