Learn More
Aviva Systems Biology Corp UTS2R Antibody - C-terminal region (ARP63549_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63549_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UTS2R. It was validated on Western Blot. . Alternate names: GPR14, UTR, UTR2, UR-2-RApplicationWB. Species reactivity: Human
Catalog No. 50-288-006