Learn More
BPS Bioscience Inc UTX (KDM6A) Homogeneous Assay Kit, 384 reactions
SDP
Catalog No. NC4036032
Encompass
The UTX (KDM6A) Homogenous Assay Kit is designed to measure activity of the UTX (Ubiquitously transcribed tetratricopeptide repeat, X chromosome, also known as KDM6A) for screening and profiling purposes. The kit comes in a convenient format 384-well AlphaLISA® format, with enough purified recombinant UTX (amino acids 1-1401), primary antibody, substrate and buffers for 400 enzyme reactions.
Catalog No. NC4036032 Supplier BPS Bioscience Inc Supplier No. 50516
The UTX (KDM6A) Homogenous Assay Kit is designed to measure activity of the UTX (Ubiquitously transcribed tetratricopeptide repeat, X chromosome, also known as KDM6A) for screening and profiling purposes. The kit comes in a convenient format 384-well AlphaLISA® format, with enough purified recombinant UTX (amino acids 1-1401), primary antibody, substrate and buffers for 400 enzyme reactions.