Learn More
Mopec UV Absorbing Goggles
SDP
Catalog No. 22446177 Shop All Mopec Products
Change view
Click to view available options
Lens Tint:
Clear
Orange
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
22-446-177 Clear
22-446-171 Yellow
22-446-122 Orange
22-446-143 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-446-177 Supplier Mopec Supplier No. FS175
Add to Cart
Add to Cart
Lightweight Anti-fog polycarbonate goggles designed to provide UV protection and reduce eye fatigue and blue haze.
  • Comfortable to wear
  • Provides 100% eye protection
  • Lightweight
  • Eliminates blue haze interference
  • Can be worn over glasses
  • Meets ANSI Z87.1 and OSHA 1910.133 standards and is designed to fit comfortably over prescription glasses.
  • Available in clear, orange, yellow, and red lenses.
Specifications
Lens Coating Anti-fog
Lens Tint Clear
Product Type UV-absorbing Goggles
Disposable Reusable
Lens Material Polycarbonate (Lightweight)
Frame Material Polycarbonate
UV Protection 100% UV
Frame Type Standard
Autoclavable Not Autoclavable