Learn More
MSC Value Collection UV Protection Bag, 8 in., 3.0 mil, 1000/Pack
Catalog No. 501920673
Encompass
The UV Protection Bag is designed to protect light-sensitive contents, measuring 8 in. wide with a thickness of 3.0 mil. Its amber color and reclosable seal make it suitable for various laboratory and industrial applications requiring light blockage and secure storage.
  • UV protection bag type
  • Width: 8 in.
  • Height: 8 in.
  • Thickness: 3.0 mil
  • Amber color
  • Reclosable
Catalog No. 50-192-0673 Supplier MSC Supplier No. 39530845
The UV Protection Bag is designed to protect light-sensitive contents, measuring 8 in. wide with a thickness of 3.0 mil. Its amber color and reclosable seal make it suitable for various laboratory and industrial applications requiring light blockage and secure storage.
  • UV protection bag type
  • Width: 8 in.
  • Height: 8 in.
  • Thickness: 3.0 mil
  • Amber color
  • Reclosable