Learn More
Med Vet International UV Blocking Medication Bag Cover 8" x 14", J1619B, 10/Pack
Catalog No. 5000583783
Encompass_Preferred
UV Blocking Medication Bags. These bags protect UV light sensitive meds from UV light. 8"W x 14"H. 10/pk.
Catalog No. 50-005-83783 Supplier Med Vet International Supplier No. JOR1619B
UV Blocking Medication Bags. These bags protect UV light sensitive meds from UV light. 8"W x 14"H. 10/pk.