Learn More
Research Products International Corp UV Chamber for PCR, 254nm, Wavelength
SDP
Catalog No. 50212396
Encompass
Innovative chamber enhances sensitive laboratory procedures such as PCR and tissue culture protocols. Unit contains a 254 nm ultraviolet lamp that effectively reduces airborne contaminants. Activating the UV lamp for 15 minutes prior to procedure and 15 minutes after procedure is completed decontaminates all interior surfaces.
Chamber is constructed of thick clear acrylic for durability and protection from beta radiation. Front viewing panel is 1/2 inch thick. One piece side and back wall are 3/8 inch thick.
Non-ventilated hood contains all electrical components and is easily removed.
Access to chamber is provided by two hinged doors that are also removable.
Built-in electronic sensor automatically turns off UV light when doors are opened. Supplied with UV and fluorescent lamps, two acrylic shelves, autoclavable tray base, power cord and one year limited warranty.
Catalog No. 50-212-396 Supplier Research Products International Corp Supplier No. 160825
Innovative chamber enhances sensitive laboratory procedures such as PCR and tissue culture protocols. Unit contains a 254 nm ultraviolet lamp that effectively reduces airborne contaminants. Activating the UV lamp for 15 minutes prior to procedure and 15 minutes after procedure is completed decontaminates all interior surfaces.
Chamber is constructed of thick clear acrylic for durability and protection from beta radiation. Front viewing panel is 1/2 inch thick. One piece side and back wall are 3/8 inch thick.
Non-ventilated hood contains all electrical components and is easily removed.
Access to chamber is provided by two hinged doors that are also removable.
Built-in electronic sensor automatically turns off UV light when doors are opened. Supplied with UV and fluorescent lamps, two acrylic shelves, autoclavable tray base, power cord and one year limited warranty.