Learn More
Alkali Scientific UV CLAVE? ULTRAVIOLET CHAMBER
SDP
Catalog No. 502388836
Encompass_Preferred
Improves quality of PCR research by reducing potential contaminants/span> Multiple lamps and reflective chamber walls provide true 360 ∞ irradiation/span> Proprietary UVC transparent shelf, eliminates potential for missed coverage (blind spots)/span> Inactivates DNA, RNA, viruses & bacteria on PCR vessels/span> Adjustable cycle time /span
Catalog No. 50-238-8836 Supplier Alkali Scientific Supplier No. RB1450
Add to Cart
Add to Cart
Improves quality of PCR research by reducing potential contaminants/span> Multiple lamps and reflective chamber walls provide true 360 ∞ irradiation/span> Proprietary UVC transparent shelf, eliminates potential for missed coverage (blind spots)/span> Inactivates DNA, RNA, viruses & bacteria on PCR vessels/span> Adjustable cycle time /span