Learn More
Medchemexpress LLC UV Cleavable Biotin-PEG2-Azide | 1192802-98-4 | 95.6% | C38H51N7O9S | 25 MG
SDP
Catalog No. 5000357121
Encompass_Preferred
UV Cleavable Biotin-PEG2-Azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent.
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAC) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-57121 Supplier Medchemexpress LLC Supplier No. 25MGHY140920
UV Cleavable Biotin-PEG2-Azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent.
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAC) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups