Learn More
Medchemexpress LLC UV cleavable biotin-PEG2-azide | 1192802-98-4 | MFCD11845531 | 95.6% | C38H51N7O9S | 10MG
SDP
Supplier:  Medchemexpress LLC HY14092010MG
Encompass_Preferred
UV Cleavable Biotin-PEG2-Azide is a photocleavable biotinylation reagent containing a PEG2 spacer and an azide group. It enables capture of biomolecules via biotin-avidin interactions and controlled release upon UV irradiation, and is compatible with copper-catalyzed and strain-promoted azide-alkyne click chemistry.
  • Photocleavable biotin enables UV-triggered release.
  • Azide functional group for CuAAC and SPAAC conjugation.
  • PEG2 spacer improves solubility and reduces steric hindrance.
  • High purity suitable for biochemical applications.
  • Available in small milligram-scale packages for research use.
Catalog No. 50-002-60469