Learn More
MSC Groz UV Germicidal Sanitizer Cabinet, 36 Safety Glasses Capacity, 15 Minute Timer, 1 ea
Catalog No. 5000688726
Encompass
This UV germicidal sanitizing cabinet is designed for the effective disinfection of safety eyewear and personal protective equipment. It utilizes UV light technology to ensure a sanitary environment, making it a reliable solution for maintaining hygiene standards in lab, shop, and industrial settings.
  • Capacity for 36 pairs of safety glasses
  • Integrated 15 minute timer for controlled operation
  • Built-in safety features
  • Fully tested for consistent performance
Catalog No. 50-006-88726 Supplier MSC Supplier No. 96313457
This UV germicidal sanitizing cabinet is designed for the effective disinfection of safety eyewear and personal protective equipment. It utilizes UV light technology to ensure a sanitary environment, making it a reliable solution for maintaining hygiene standards in lab, shop, and industrial settings.
  • Capacity for 36 pairs of safety glasses
  • Integrated 15 minute timer for controlled operation
  • Built-in safety features
  • Fully tested for consistent performance