Learn More
Molecular Dimensions UV HANGING DROP SHEETS (100)
SDP
Catalog No. 501038531
Encompass
UV HANGING DROP SHEETS (100) Hanging-drop sheets with 96 round wells for easy positioning of crystallization drops for hanging-drop experiments.
Catalog No. 50-103-8531 Supplier Molecular Dimensions Supplier No. MD1100HD
UV HANGING DROP SHEETS (100) Hanging-drop sheets with 96 round wells for easy positioning of crystallization drops for hanging-drop experiments.