Learn More
Grainger Elga UV Lamp, 185 to 254 nm, 13 in. Bulb, 1 ea.
Catalog No. 502790083
Encompass_Preferred
This UV lamp is designed for use with PURELAB water purification systems, providing effective irradiation at specific wavelengths to support water treatment processes.
  • Wavelength range: 185 nm to 254 nm
  • Compatible with Purelab Flex series including models PF1XXXXM2, PF1XXXXM3, and PF1XXXXM4
  • Bulb length: 13 in.
Catalog No. 50-279-0083 Supplier Grainger Supplier No. 16D273
This UV lamp is designed for use with PURELAB water purification systems, providing effective irradiation at specific wavelengths to support water treatment processes.
  • Wavelength range: 185 nm to 254 nm
  • Compatible with Purelab Flex series including models PF1XXXXM2, PF1XXXXM3, and PF1XXXXM4
  • Bulb length: 13 in.