Learn More
EZKEM UV LAMP for CN Analysis 8W 312NM 12 INCHES
SDP
Catalog No. NC1813043
Encompass_Preferred
EZkem’s 12 inch, 8 watt, 312 nm UV Lamp is used on Alpkem and OI Analytical autoanalyzers, cyanide solution, FS3000, FSIII, FSIV – FIA and SFA flow analyzers.
Catalog No. NC1813043 Supplier EZKEM Supplier No. A001682
EZkem’s 12 inch, 8 watt, 312 nm UV Lamp is used on Alpkem and OI Analytical autoanalyzers, cyanide solution, FS3000, FSIII, FSIV – FIA and SFA flow analyzers.