Learn More
Perkin Elmer US LLC UV Lamp Complete - ELP
Catalog No. 502112079
Encompass_Preferred
For legacy LAMBDA spectrometers, PerkinElmer supplies reliable, long lasting deuterium and tungsten-halogen light sources. We recommend these replacement bulbs be installed by a PerkinElmer Service Engineer since optimal spectrometer performance depends upon the lamps precise alignment.
This item is not returnable. View return policy
Catalog No. 50-211-2079 Supplier Perkin Elmer US LLC Supplier No. L6022728
For legacy LAMBDA spectrometers, PerkinElmer supplies reliable, long lasting deuterium and tungsten-halogen light sources. We recommend these replacement bulbs be installed by a PerkinElmer Service Engineer since optimal spectrometer performance depends upon the lamps precise alignment.