Learn More
Cole-Parmer Handheld UV Lamp, 6 Watt, 254/365 nm, AC Operated; 110 VAC, 50/60 Hz
Catalog No. NC1903745 Shop All Cole-Parmer Products
Encompass_Preferred
The series of lightweight, handheld lamps offer UV wavelengths of 365 and 254 nm, or a combination of the two. They’re cordless and use 12 VDC rechargeable batteries. They can sustain three hours of continuous use or about 500 intermittent uses when fully charged. The models equipped with an automatic safety switch prevent harmful exposure to UV radiation if the lamp is turned upside down.
Catalog No. NC1903745 Supplier Cole-Parmer Supplier No. 0981807
The series of lightweight, handheld lamps offer UV wavelengths of 365 and 254 nm, or a combination of the two. They’re cordless and use 12 VDC rechargeable batteries. They can sustain three hours of continuous use or about 500 intermittent uses when fully charged. The models equipped with an automatic safety switch prevent harmful exposure to UV radiation if the lamp is turned upside down.