Learn More
Ace Glass, Inc. UV lamp, immersion, 550 watt, 109.54mm arc length, 236.54mmoverall length, metal-ended, no insulation, NEMA 15P power cord included
SDP
Catalog No. 50975483
Encompass
550watt med. pressure, quartz mercury-vapor lamp, for use in all ACE immersion wells. Arc length: 109.54mm. Overall length: 236.54mm. Supplied with 61cm PTFE covered lead wires, fitted with pin type connectors, and 6 cord to power supply.
Catalog No. 50-975-483 Supplier Ace Glass, Inc. Supplier No. 782536
550watt med. pressure, quartz mercury-vapor lamp, for use in all ACE immersion wells. Arc length: 109.54mm. Overall length: 236.54mm. Supplied with 61cm PTFE covered lead wires, fitted with pin type connectors, and 6 cord to power supply.