Learn More
Hach Company Ultraviolet Lamp Kit, 230 Vac, 50Hz
Catalog No. 501996583 Shop All Hach Company Products
Encompass
Contains mercury. Dispose of according to local, state and federal regulations. Shortwave (254 nm) UV lamps for field or laboratory use. Either lamp can be used in the lab with power supply while Portable Field Pack can only be used with the 1.75 lamp. Portable Field Pack requires a 9-Volt battery (not included) and has a belt clip for easy traveling. Both lamps are physically identical except for bulb and cord length (this is the shorter 1.75 version). Kit Includes UV Goggles, 220Vac Power Supply and 1.75" UV Lamp. NOTE: Not for use in microbiological applications. UV Lamp contains mercury. Dispose according to state, local or federal regulations.
Catalog No. 50-199-6583 Supplier Hach Company Supplier No. 2704503
Contains mercury. Dispose of according to local, state and federal regulations. Shortwave (254 nm) UV lamps for field or laboratory use. Either lamp can be used in the lab with power supply while Portable Field Pack can only be used with the 1.75 lamp. Portable Field Pack requires a 9-Volt battery (not included) and has a belt clip for easy traveling. Both lamps are physically identical except for bulb and cord length (this is the shorter 1.75 version). Kit Includes UV Goggles, 220Vac Power Supply and 1.75" UV Lamp. NOTE: Not for use in microbiological applications. UV Lamp contains mercury. Dispose according to state, local or federal regulations.