Learn More
ERA WATERS CORP UV LAMP /PAT 700
SDP
Catalog No. NC1723617
Encompass
  • Critical replacement component for TOC Analyzer
  • Ensures efficient breakdown of organic molecules in water samples
  • Supports compliance with global pharmacopeia standards
  • Monitors lamp efficiency for reliability
  • Lightweight design at 0.485 lbs
  • Available with a lead time of 1 business day
  • Suitable for ambient storage conditions
  • Designed for pharmaceutical applications
  • Single component per package
  • Used in process water consumables
Catalog No. NC1723617 Supplier ERA WATERS CORP Supplier No. 20037
  • Critical replacement component for TOC Analyzer
  • Ensures efficient breakdown of organic molecules in water samples
  • Supports compliance with global pharmacopeia standards
  • Monitors lamp efficiency for reliability
  • Lightweight design at 0.485 lbs
  • Available with a lead time of 1 business day
  • Suitable for ambient storage conditions
  • Designed for pharmaceutical applications
  • Single component per package
  • Used in process water consumables