Learn More
Hach Company Lamp, Ultraviolet, Portable
Supplier:  Hach Company 2415200
Encompass
Hand-held, battery-operated, long-wave UV lamp 365 nm. Uses for AA batteries not included (For Microbiology Applications). 4 W. Note: Not for digestion.
Catalog No. 50-199-5951