Learn More
VEOLIA WTS UV Lamp, Replacement
Catalog No. NC0945889
Encompass
UV Lamp for the Sievers M-Series; Lifetime of lamp is 6 months of operation.
Catalog No. NC0945889 Supplier VEOLIA WTS Supplier No. ARK 3501001
UV Lamp for the Sievers M-Series; Lifetime of lamp is 6 months of operation.