Learn More
Perkin Elmer US LLC UV Light Shield for NexION 350
Catalog No. 502112029
Encompass_Preferred
The UV light shield protects users from rays given off by the plasma in the instrument and secures the spray chamber to the base of the injector. It is included in the Single Cell Kit for NexION 350 (N8140032).
Catalog No. 50-211-2029 Supplier Perkin Elmer US LLC Supplier No. N8142047
The UV light shield protects users from rays given off by the plasma in the instrument and secures the spray chamber to the base of the injector. It is included in the Single Cell Kit for NexION 350 (N8140032).