Learn More
Perkin Elmer US LLC UV Material Disposable Macro Cells - 10 mm, Qty. 100
Catalog No. 502112092
Encompass_Preferred
These Disposable Macro Cells are made of UV material, which has a useful spectral range of 220-900 nm. They have a volume of 2.5-4.5 mL, a path length of 10 mm, and outside dimensions of 45 mm (H) x 12.5 mm (W) x 12.5 mm (D). 100 per package.
Catalog No. 50-211-2092 Supplier Perkin Elmer US LLC Supplier No. L7110269
These Disposable Macro Cells are made of UV material, which has a useful spectral range of 220-900 nm. They have a volume of 2.5-4.5 mL, a path length of 10 mm, and outside dimensions of 45 mm (H) x 12.5 mm (W) x 12.5 mm (D). 100 per package.