Learn More
Electron Microscopy Sciences UV Mounting Medium 100 ML
SDP
Catalog No. 5029580
Encompass_Preferred
Mounting medium that is designed to match the refractive index of JB-4 embedded sections. Cures within 2 minutes when exposed to long wave UV light (365nm) when applied to moist sections. Sections are mounted after last staining water rinse. Slides must be coverslipped.
Catalog No. 50-295-80 Supplier Electron Microscopy Sciences Supplier No. 17996
Mounting medium that is designed to match the refractive index of JB-4 embedded sections. Cures within 2 minutes when exposed to long wave UV light (365nm) when applied to moist sections. Sections are mounted after last staining water rinse. Slides must be coverslipped.