Learn More
Azzota Corp Uv Neutral Density Filter Set For Spectrophotometer Photometric Accuracy Calibration, Has Two Neutral Density Filters, The Testing Photometric Are 235Nm, 257Nm, 313Nm, And 350Nm
Catalog No. 507532923
Encompass_Preferred
UV neutral density filter set for spectrophotometer photometric accuracy calibration, has two Neutral Density Filters, the testing photometric are 235nm, 257nm, 313nm, and 350nm
Catalog No. 50-753-2923 Supplier Azzota Corp Supplier No. Q806
UV neutral density filter set for spectrophotometer photometric accuracy calibration, has two Neutral Density Filters, the testing photometric are 235nm, 257nm, 313nm, and 350nm