Learn More
Perkin Elmer US LLC UV Quartz SUPRASIL Rectangular Micro Cell with PTFE Lid - 10 mm, Qty. 2
Catalog No. 502112640
Encompass_Preferred
Micro cells have the same outside dimension as Macro cells. Their cell walls have been thickened, limiting the interior sample chamber width to 2 mm. Each cell has a standard path length of 10 mm with black side walls and base. Each package contains two cells. UV Quartz SUPRASIL is a quartz of the highest purity and homogeneity. An empty UV Quartz SUPRASIL cell gives a transmission of more that 80% over a spectral range of 200 - 2,500 nm. These cells are identified by a blue PerkinElmer logo.
Catalog No. 50-211-2640 Supplier Perkin Elmer US LLC Supplier No. B0631071
Micro cells have the same outside dimension as Macro cells. Their cell walls have been thickened, limiting the interior sample chamber width to 2 mm. Each cell has a standard path length of 10 mm with black side walls and base. Each package contains two cells. UV Quartz SUPRASIL is a quartz of the highest purity and homogeneity. An empty UV Quartz SUPRASIL cell gives a transmission of more that 80% over a spectral range of 200 - 2,500 nm. These cells are identified by a blue PerkinElmer logo.