Learn More
Perkin Elmer US LLC UV Shield Adapter for NexION 1000/2000
Catalog No. 502113118
Encompass_Preferred
This adapter is for use with NexION 1000 and 2000 ICP-MS series instruments. It is designed to secure the spray chamber to the base of the injector when using a UV shield.
Catalog No. 50-211-3118 Supplier Perkin Elmer US LLC Supplier No. W1037267
This adapter is for use with NexION 1000 and 2000 ICP-MS series instruments. It is designed to secure the spray chamber to the base of the injector when using a UV shield.