Learn More
Greiner Bio-One UV-Star™ 96-well Microplates
GREENER_CHOICE
Catalog No. 07000629 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
07-000-629 Black
1 options
Catalog No. 07-000-629 Supplier Greiner Bio-One Supplier No. 655809
Add to Cart
Edge
Add to Cart
Ideal for measurements of DNA / RNA and protein concentrations at 260 and 280nm, respectively. Greiner Bio-One UV-Star™ 96-Well Microplates are eliminate the use of expensive and fragile quartz glass for routine laboratory work.
  • Coated with μClear film well bottom to ensure low background for more accurate measurements
  • Innovative molding technique eliminates possibility of film detachment
  • Supplied without lid
  • Chimney-well design helps reduce cross-contamination
  • DNA measurement at 260/280nm
  • Protein measurement at 280nm
Service Plan
No
Warranty and Services
No
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Packaging Inner bags of 10
Binding Type Non Binding
Color Black
Product Type UV Microplate
Material Cycloolefin Copolymer
Volume (Metric) Well 25 to 340 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Untreated
Total Volume (Metric) Well 392 μL
Well Shape Flat Bottom (F-well)
No. per Case 40
No. per Pack 10
Quantity 40/Cs.
Well Color Black
Dimensions (L x W x H) 127.8 x 85.5 x 14.4 mm