Learn More
Microsolv Technology Corporation UV-vis Calibration Test Kit, Complete. Includes NIST Traceable Solutions, Software. Chemical Solutions Brand. 1 EA.
Catalog No. 502269768
Encompass_Preferred
This UV-vis Calibration PQ Kit™ is a series of NIST traceable solutions with easy data recording and reporting of the actual data with acceptance criteria. This will allow you to Qualify a UV-vis Spectrophotometer easily and fully in regulated laboratories. In-house Performance Qualification (PQ) of these instruments is easy and saves money and time.UV-vis Kit Includes: Absorbance Solutions: Reference Solution Diluent Wavelength Calibration Solution Stray Light Solution Software: Excel Template for Data Recording and Analysis Certificate of Analysis. Solutions will be supplied with at least six months until the expiration date. They are designed to be used promptly and often. Expiration date is two years after the date of manufacture.
This item is not returnable. View return policy
Catalog No. 50-226-9768 Supplier Microsolv Technology Corporation Supplier No. 4911501
This UV-vis Calibration PQ Kit™ is a series of NIST traceable solutions with easy data recording and reporting of the actual data with acceptance criteria. This will allow you to Qualify a UV-vis Spectrophotometer easily and fully in regulated laboratories. In-house Performance Qualification (PQ) of these instruments is easy and saves money and time.UV-vis Kit Includes: Absorbance Solutions: Reference Solution Diluent Wavelength Calibration Solution Stray Light Solution Software: Excel Template for Data Recording and Analysis Certificate of Analysis. Solutions will be supplied with at least six months until the expiration date. They are designed to be used promptly and often. Expiration date is two years after the date of manufacture.