Learn More
HEPATOCHEM PhotoRedOx Box UV-Vis Photoreactor Holder 8x 8ml vial
Catalog No. NC3714382
Encompass
Holder 8x 8ml vial for PhotoRedOx Box UV-Vis Photoreactor including an IR probe to measure temperature in a reaction vial
Catalog No. NC3714382 Supplier HEPATOCHEM Supplier No. HCK102201003
Holder 8x 8ml vial for PhotoRedOx Box UV-Vis Photoreactor including an IR probe to measure temperature in a reaction vial