Learn More
Azzota Corp Single Beam Uv-Vis Scanning Spectrophotometer. 4Nm Bandwidth, 190-1100Nm, 320 X 240 Lcd. Auto Set Wl. Usb Connection. Display/Save 200 Standard Curves/Data. W/ Kinetics Function. With Pc Software
Catalog No. 501612948
Encompass_Preferred
Single beam UV-Vis scanning spectrophotometer. 4nm bandwidth, 190-1100nm, 320 x 240 LCD. Automatically set WL. USB connection. Can display and save 200 standard curves and 200 groups of data. With Kinetics Function. with PC software
Catalog No. 50-161-2948 Supplier Azzota Corp Supplier No. SE3000PC
Add to Cart
Add to Cart
Request a Quote
Single beam UV-Vis scanning spectrophotometer. 4nm bandwidth, 190-1100nm, 320 x 240 LCD. Automatically set WL. USB connection. Can display and save 200 standard curves and 200 groups of data. With Kinetics Function. with PC software