Learn More
Azzota Corp Single Beam Uv-Vis Spectrophotometer, Wavelength 190-1100Nm, 2Nm Bandwidth, 320 X 240 Lcd. Large Screen, High Accuracy. Intelligent Type.
Catalog No. 507532966
Encompass_Preferred
Single Beam UV-VIS Spectrophotometer, Wavelength 190-1100nm, 2nm Bandwidth, 320 x 240 LCD. Large screen, high accuracy. Intelligent type.
Catalog No. 50-753-2966 Supplier Azzota Corp Supplier No. SM1800
Add to Cart
Add to Cart
Request a Quote
Single Beam UV-VIS Spectrophotometer, Wavelength 190-1100nm, 2nm Bandwidth, 320 x 240 LCD. Large screen, high accuracy. Intelligent type.