Learn More
Azzota Corp Uv-Visible Spectrophotometer, 2Nm Bandwidth, 190-1100Nm, 128X64 Lcd. Auto Set Wl. Usb Connection. Can Display And Save 200 Standard Curves And 200 Groups Of Data. Pc Scanning Software Included.
Catalog No. 501613001
Encompass_Preferred
UV-Visible Spectrophotometer, 2nm bandwidth, 190-1100nm, 128x64 LCD. Automatically set WL. USB connection. Can display and save 200 standard curves and 200 groups of data. PC scanning software included.
Catalog No. 50-161-3001 Supplier Azzota Corp Supplier No. SM1800PC
UV-Visible Spectrophotometer, 2nm bandwidth, 190-1100nm, 128x64 LCD. Automatically set WL. USB connection. Can display and save 200 standard curves and 200 groups of data. PC scanning software included.