Learn More
Azzota Corp Uv-Visible Spectrophotometer, 4Nm Bandwidth, 190-1100Nm, 128 X 64 Lcd. Automatically Set Wl. Usb Connection. Display/Save 200 Standard Curves/Data. W/Kinetics Function. Pc Wl Scan Software Included.
Catalog No. 501613000
Encompass_Preferred
UV-Visible Spectrophotometer, 4nm bandwidth, 190-1100nm, 128 x 64 LCD. Automatically set WL. USB connection. Can display and save 200 standard curves and 200 groups of data. With Kinetics Function. PC WL scan software included.
Catalog No. 50-161-3000 Supplier Azzota Corp Supplier No. SM1600PC
Add to Cart
Add to Cart
Request a Quote
UV-Visible Spectrophotometer, 4nm bandwidth, 190-1100nm, 128 x 64 LCD. Automatically set WL. USB connection. Can display and save 200 standard curves and 200 groups of data. With Kinetics Function. PC WL scan software included.