Learn More
Perkin Elmer US LLC UV Wavelength Calibration Kit
Catalog No. 502117272
Encompass_Preferred
This kit includes Part Number Description Content Matix Size N9304162 UV Wavelength Calibration Standard 5 µg/mL:Ca Hydrocarbon Oil 100g 100 µg/mL: As, La, Li, Mn, Mo, Ni, Na 500 µg/mL: K, P 100 µg/mL: As, La, Li, Mn, Mo, Ni, Na N9308255 STD - Scandium 100 µg/mL:Sc Hydrocarbon Oil 10g
Catalog No. 50-211-7272 Supplier Perkin Elmer US LLC Supplier No. N9304163
This kit includes Part Number Description Content Matix Size N9304162 UV Wavelength Calibration Standard 5 µg/mL:Ca Hydrocarbon Oil 100g 100 µg/mL: As, La, Li, Mn, Mo, Ni, Na 500 µg/mL: K, P 100 µg/mL: As, La, Li, Mn, Mo, Ni, Na N9308255 STD - Scandium 100 µg/mL:Sc Hydrocarbon Oil 10g