Learn More
Research Products International Corp UV/White Light Converter, 21x26cm
SDP
Catalog No. 501977167
Encompass
Unique design concept converts UV radiation from any transilluminator to white light. Ideal for viewing protein gels, and coomassie blue or silver stained media. Converter assembly has a metal housing with convenient handles for easy lifting. Transilluminator in photo available separately.
Catalog No. 50-197-7167 Supplier Research Products International Corp Supplier No. 380191
Add to Cart
Add to Cart
Unique design concept converts UV radiation from any transilluminator to white light. Ideal for viewing protein gels, and coomassie blue or silver stained media. Converter assembly has a metal housing with convenient handles for easy lifting. Transilluminator in photo available separately.