Learn More
Research Products International Corp UV/White Light Converter, 20 x 40cm
SDP
Catalog No. 50212711
Encompass
Unique design concept converts UV radiation from any transilluminator to white light. Ideal for viewing protein gels, and coomassie blue or silver stained media. Converter assembly has a metal housing with convenient handles for easy lifting. Transilluminator in photo available separately.
Catalog No. 50-212-711 Supplier Research Products International Corp Supplier No. 380101
Unique design concept converts UV radiation from any transilluminator to white light. Ideal for viewing protein gels, and coomassie blue or silver stained media. Converter assembly has a metal housing with convenient handles for easy lifting. Transilluminator in photo available separately.