Learn More
Perkin Elmer US LLC UV WinLab Enhanced Security Software upgrade for UV/Vis
Catalog No. 502016623
Encompass_Preferred
UV WinLab Enhanced Security is the instrument control and data manipulation software for the latest range of PerkinElmer LAMBDA 365, 650, 750, 850, 950, 1050. High Performance UV/Vis and UV/Vis/NIR spectrophotometers. Designed for busy pharmaceutical, manufacturing, industrial, food, or academic laboratories that require efficient operation combined with a wide-range of capabilities, this comprehensive package is designed for simplicity and efficiency in data collection, processing and generating results. Our UV WinLab Enhanced Security software upgrade is: Windows 10 enabled Compatible with Lambda 365 instruments This upgrade can be used with UV WinLab ES V6.0.4 or later. If you have a previous version software this upgrade will not work properly. Please see L6100126.
Catalog No. 50-201-6623 Supplier Perkin Elmer US LLC Supplier No. L6100155
UV WinLab Enhanced Security is the instrument control and data manipulation software for the latest range of PerkinElmer LAMBDA 365, 650, 750, 850, 950, 1050. High Performance UV/Vis and UV/Vis/NIR spectrophotometers. Designed for busy pharmaceutical, manufacturing, industrial, food, or academic laboratories that require efficient operation combined with a wide-range of capabilities, this comprehensive package is designed for simplicity and efficiency in data collection, processing and generating results. Our UV WinLab Enhanced Security software upgrade is: Windows 10 enabled Compatible with Lambda 365 instruments This upgrade can be used with UV WinLab ES V6.0.4 or later. If you have a previous version software this upgrade will not work properly. Please see L6100126.