Learn More
Perkin Elmer US LLC UV WinLab Software for Uv/Vis
Catalog No. 502016627
Encompass_Preferred
UV WinLab is the instrument control and data manipulation software for the latest range of PerkinElmer LAMBDA 365, 650, 750, 850, 950, 1050. High Performance UV/Vis and UV/Vis/NIR spectrophotometers. Designed for busy pharmaceutical, manufacturing, industrial, food, or academic laboratories that require efficient operation combined with a wide-range of capabilities, this comprehensive package is designed for simplicity and efficiency in data collection, processing and generating results. With our UV WinLab Enhanced Security software you can: Focus on the results, not the tasks Take control of your instrument Automatically configure accessories Customize your workbench Stay compliant with 21 CFR Part 11 UV WinLab provides security without compromising laboratory productivity.
Catalog No. 50-201-6627 Supplier Perkin Elmer US LLC Supplier No. L6100127
UV WinLab is the instrument control and data manipulation software for the latest range of PerkinElmer LAMBDA 365, 650, 750, 850, 950, 1050. High Performance UV/Vis and UV/Vis/NIR spectrophotometers. Designed for busy pharmaceutical, manufacturing, industrial, food, or academic laboratories that require efficient operation combined with a wide-range of capabilities, this comprehensive package is designed for simplicity and efficiency in data collection, processing and generating results. With our UV WinLab Enhanced Security software you can: Focus on the results, not the tasks Take control of your instrument Automatically configure accessories Customize your workbench Stay compliant with 21 CFR Part 11 UV WinLab provides security without compromising laboratory productivity.