Learn More
Perkin Elmer US LLC UV WinLab Software Upgrade for UV/Vis
Catalog No. 502016622
Encompass_Preferred
UV Winlab Software upgrade is the instrument control and data manipulation software for the latest range of PerkinElmer LAMBDA 365, 650, 750, 850, 950, 1050. High Performance UV/Vis and UV/Vis/NIR spectrophotometers. Designed for busy pharmaceutical, manufacturing, industrial, food, or academic laboratories that require efficient operation combined with a wide-range of capabilities, this comprehensive package is designed for simplicity and efficiency in data collection, processing, and generating results. This kit includes the upgrade software, an upgrade manual and a license for one user. Our UV WinLab standard software upgrade is: Windows 10 enabled Compatible with LAMBDA 365 instruments This upgrade can be used with UV WinLab V6.0.4 or later. If you have an earlier version of this software, this upgrade will not work properly. Please see UV WinLab v7.1 Standard Software (L6100127).
Catalog No. 50-201-6622 Supplier Perkin Elmer US LLC Supplier No. L6100154
UV Winlab Software upgrade is the instrument control and data manipulation software for the latest range of PerkinElmer LAMBDA 365, 650, 750, 850, 950, 1050. High Performance UV/Vis and UV/Vis/NIR spectrophotometers. Designed for busy pharmaceutical, manufacturing, industrial, food, or academic laboratories that require efficient operation combined with a wide-range of capabilities, this comprehensive package is designed for simplicity and efficiency in data collection, processing, and generating results. This kit includes the upgrade software, an upgrade manual and a license for one user. Our UV WinLab standard software upgrade is: Windows 10 enabled Compatible with LAMBDA 365 instruments This upgrade can be used with UV WinLab V6.0.4 or later. If you have an earlier version of this software, this upgrade will not work properly. Please see UV WinLab v7.1 Standard Software (L6100127).