Learn More
Med Alliance Group Inc UVC 254 TRI CHECK, POUCH
SDP
Catalog No. NC2232058
Encompass
254 TRI Check. Using a patented, photochromic ink, TRI Check dosimeters react to UV-C at 254 nm.25 Dosimeters per Pouch.
This item is not returnable. View return policy
Catalog No. NC2232058 Supplier Med Alliance Group Inc Supplier No. UVC254TCK POUCH
254 TRI Check. Using a patented, photochromic ink, TRI Check dosimeters react to UV-C at 254 nm.25 Dosimeters per Pouch.